Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate 95%

Supplier: Apollo Scientific

OR930286-1G OR930286-5G
APOSOR930286-1GEA 190 EUR
APOSOR930286-1G APOSOR930286-5G
Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate 95%
Ethyl-2-nitrilo-3-(2-furyl)prop-2-enoate
Formula: C₁₀H₉NO₃
MW: 191,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137603
CAS Number: 23973-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR