4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 95%

Supplier: Apollo Scientific

Synonyms: TEMPONE, 4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 4-Oxo-TEMPO

OR930328-25G OR930328-5G
APOSOR930328-25GEA 931 EUR
APOSOR930328-25G APOSOR930328-5G
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 95%
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl
Formula: C₉H₁₆NO₂
MW: 170,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006187
CAS Number: 2896-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR