2,3,5-Trichlorophenol 95%

Supplier: Apollo Scientific

OR930342-1G OR930342-5G
APOSOR930342-1GEA 292 EUR
APOSOR930342-1G APOSOR930342-5G
2,3,5-Trichlorophenol 95%
2,3,5-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002164
CAS Number: 933-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR