3-(4-Chlorophenyl)propiolic acid 95%
Supplier: Apollo Scientific
Synonyms:
3-(4-Chlorophenyl)-2-propynoic acid, (4-Chlorophenyl)propargylic acid
OR930388-1G
OR930388-250MG
APOSOR930388-1GEA
66.7
EUR
APOSOR930388-1G
APOSOR930388-250MG
3-(4-Chlorophenyl)propiolic acid 95%
3-(4-Chlorophenyl)propiolic acid
Formula:
C₉H₅ClO₂ MW: 180.5878 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046550 CAS Number: 3240-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...