1-Phenyl-2-pyrrolidone 98%

Supplier: Apollo Scientific

Synonyms: 1-Phenyl-2-pyrrolidinone

OR930513-100G
APOSOR930513-100GEA 85.7 EUR
APOSOR930513-100G
1-Phenyl-2-pyrrolidone 98%
1-Phenyl-2-pyrrolidone
Formula: C₁₀H₁₁NO
MW: 161,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003192
CAS Number: 4641-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR