N-[3-(Dimethylamino)propyl]acrylamide 96%

Supplier: Apollo Scientific

OR930552-25G OR930552-100G
APOSOR930552-25GEA 61.1 EUR
APOSOR930552-25G APOSOR930552-100G
N-[3-(Dimethylamino)propyl]acrylamide 96%
N-[3-(Dimethylamino)propyl]acrylamide
Formula: C₈H₁₆N₂O
MW: 156,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059902
CAS Number: 3845-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR