5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR930969-1G
OR930969-250MG
APOSOR930969-1GEA
81.2
EUR
APOSOR930969-1G
APOSOR930969-250MG
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95%
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Formula:
C₈H₅ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD12962789 CAS Number: 954112-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...