2-Benzylphenol 98%
Supplier: Apollo Scientific
OR931049-100G
OR931049-500G
APOSOR931049-100GEA
0
EUR
APOSOR931049-100G
APOSOR931049-500G
2-Benzylphenol 98%
2-Benzylphenol
Formula:
C₁₃H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD00002242 CAS Number: 28994-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...