Dihydrolinalool 95%

Supplier: Apollo Scientific

Synonyms: 3,7-Dimethyl-6-octen-3-ol, 3,7-Dimethyloct-6-en-3-ol

OR931110-25G OR931110-100G
APOSOR931110-25GEA 139 EUR
APOSOR931110-25G APOSOR931110-100G
Dihydrolinalool 95%
Dihydrolinalool
Formula: C₁₀H₂₀O
MW: 156,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059140
CAS Number: 18479-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR