2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol 97%

Supplier: Apollo Scientific

Synonyms: 2-[[tert-Butyl(dimethyl)silyl]oxy]ethan-1-ol, 2-((tert-Butyldimethylsilyl)oxy)ethanol

OR931367-100G OR931367-10G OR931367-25G
APOSOR931367-100GEA 865 EUR
APOSOR931367-100G APOSOR931367-10G APOSOR931367-25G
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol 97%
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol
Formula: C₈H₂₀O₂Si
MW: 176,33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261150
CAS Number: 102229-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR