2-Butylimidazole 95%
Supplier: Apollo Scientific
OR931417-25G
OR931417-5G
APOSOR931417-25GEA
121
EUR
APOSOR931417-25G
APOSOR931417-5G
2-Butylimidazole 95%
2-Butylimidazole
Formula:
C₇H₁₂N₂ MW: 124,1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792664 CAS Number: 50790-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...