2-(4-Methylphenyl)ethanol 95%

Supplier: Apollo Scientific

Synonyms: 2-(p-Tolyl)ethanol

OR931463-25G
APOSOR931463-25GEA 58 EUR
APOSOR931463-25G
2-(4-Methylphenyl)ethanol 95%
2-(4-Methylphenyl)ethanol
Formula: C₉H₁₂O
MW: 136,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002904
CAS Number: 699-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR