(E)-4-Phenylbut-3-enoic acid 98%

Supplier: Apollo Scientific

OR931557-10G OR931557-1G OR931557-25G
APOSOR931557-10GEA 234 EUR
APOSOR931557-10G APOSOR931557-1G APOSOR931557-25G
(E)-4-Phenylbut-3-enoic acid 98%
(E)-4-Phenylbut-3-enoic acid
Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00002783
CAS Number: 1914-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR