4-Amino-3-phenylbutyric acid hydrochloride 98%

Supplier: Apollo Scientific

Synonyms: 4-Amino-3-phenylbutanoic acid

OR931735-10G OR931735-5G
APOSOR931735-10GEA 190 EUR
APOSOR931735-10G APOSOR931735-5G
4-Amino-3-phenylbutyric acid hydrochloride 98%
4-Amino-3-phenylbutyric acid hydrochloride
Formula: C₁₀H₁₄ClNO₂
MW: 215,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075860
CAS Number: 1078-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR