(4-Methoxyphenyl)acetaldehyde 95%

Supplier: Apollo Scientific

Synonyms: 2-(4-Methoxyphenyl)acetaldehyde

OR932105-1G OR932105-250MG
APOSOR932105-1GEA 181 EUR
APOSOR932105-1G APOSOR932105-250MG
(4-Methoxyphenyl)acetaldehyde 95%
(4-Methoxyphenyl)acetaldehyde
Formula: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02261769
CAS Number: 5703-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR