m-Tolyl acetate 95%

Supplier: Apollo Scientific

OR932170-100G OR932170-25G
APOSOR932170-100GEA 299 EUR
APOSOR932170-100G APOSOR932170-25G
m-Tolyl acetate 95%
m-Tolyl acetate
Formula: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041910
CAS Number: 122-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR