2-Bromoterephthalic acid 98+%
Supplier: Apollo Scientific
|
Warning
|
OR9323-100G
OR9323-25G
OR9323-5G
APOSOR9323-100GEA
139
EUR
APOSOR9323-100G
APOSOR9323-25G
APOSOR9323-5G
2-Bromoterephthalic acid 98+%
2-Bromoterephthalic acid
Formula:
C₈H₅BrO₄ Melting Pt: 295…297 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002403 CAS Number: 586-35-6 EINECS: 209-572-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...