2-tert-Butyl-1,4-benzoquinone 98%
Supplier: Apollo Scientific
OR932648-25G
OR932648-5G
APOSOR932648-25GEA
74.2
EUR
APOSOR932648-25G
APOSOR932648-5G
2-tert-Butyl-1,4-benzoquinone 98%
2-tert-Butyl-1,4-benzoquinone
Formula:
C₁₀H₁₂O₂ MW: 164,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00666928 CAS Number: 3602-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...