2-Amino-4,5-dimethoxybenzonitrile
Supplier: Apollo Scientific
|
Warning
|
OR9328-5G
APOSOR9328-5GEA
65.4
EUR
APOSOR9328-5G
2-Amino-4,5-dimethoxybenzonitrile
2-Amino-4,5-dimethoxybenzonitrile
Formula:
C₉H₁₀N₂O₂ Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058682 CAS Number: 26961-27-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...