1-(tert-Butyldimethylsilyl)imidazole 95%
Supplier: Apollo Scientific
Synonyms:
1-(tert-Butyldimethylsilyl)-1H-imidazole
OR932834-1G
OR932834-5G
OR932834-25G
APOSOR932834-1GEA
80
EUR
APOSOR932834-1G
APOSOR932834-5G
APOSOR932834-25G
1-(tert-Butyldimethylsilyl)imidazole 95%
1-(tert-Butyldimethylsilyl)imidazole
Formula:
C₉H₁₈N₂Si MW: 182,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011682 CAS Number: 54925-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...