Tetramethyl-1,3-cyclobutanedione

Supplier: Apollo Scientific

Synonyms: Tetramethylcyclobutane-1,3-dione

OR933088-100G OR933088-25G OR933088-50G
APOSOR933088-100GEA 522 EUR
APOSOR933088-100G APOSOR933088-25G APOSOR933088-50G
Tetramethyl-1,3-cyclobutanedione
Tetramethyl-1,3-cyclobutanedione
Formula: C₈H₁₂O₂
MW: 140,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001331
CAS Number: 933-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR