2-Amino-4-methylthiophene hydrochloride 95%

Supplier: Apollo Scientific

Synonyms: 4-Methylthiophen-2-amine hydrochloride

OR933181-1G OR933181-250MG
APOSOR933181-1GEA 807 EUR
APOSOR933181-1G APOSOR933181-250MG
2-Amino-4-methylthiophene hydrochloride 95%
2-Amino-4-methylthiophene hydrochloride
Formula: C₅H₈ClNS
MW: 149,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21604144
CAS Number: 930299-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR