2-(Ammoniooxy)propane chloride 98%

Supplier: Apollo Scientific
Danger

Synonyms: O-propan-2-ylhydroxylamine hydrochloride, O-Isopropylhydroxylamine hydrochloride

OR933332-10G OR933332-1G OR933332-5G
APOSOR933332-10GEA 945 EUR
APOSOR933332-10G APOSOR933332-1G APOSOR933332-5G
2-(Ammoniooxy)propane chloride 98%
2-(Ammoniooxy)propane chloride
Formula: C₃H₁₀ClNO
MW: 111,5706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00231457
CAS Number: 4490-81-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR