4-Bromomethyl-6,7-dimethoxycoumarin
Supplier: Apollo Scientific
|
Danger
|
OR9344-1G
OR9344-250MG
APOSOR9344-1GEA
219
EUR
APOSOR9344-1G
APOSOR9344-250MG
4-Bromomethyl-6,7-dimethoxycoumarin
4-Bromomethyl-6,7-dimethoxycoumarin
Formula:
C₁₂H₁₁BrO₄ MW: 299,12 g/mol Melting Pt: 214…216 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011570 CAS Number: 88404-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...