Dibenzothiophene-4-boronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Dibenzothiopheneboronic acid

OR9356-100G OR9356-25G OR9356-5G
APOSOR9356-100GEA 228 EUR
APOSOR9356-100G APOSOR9356-25G APOSOR9356-5G
Dibenzothiophene-4-boronic acid 97%
Dibenzothiophene-4-boronic acid
Formula: C₁₂H₉BO₂S
MW: 228,08 g/mol
Melting Pt: 327…330 °C
Storage Temperature: Ambient
MDL Number: MFCD01318182
CAS Number: 108847-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR