2-Chloro-3-pyridineboronic acid 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Chloro-3-pyridinylboronic acid, (2-Chloropyridin-3-yl)boronic acid, 2-Chloropyridine-3-boronic acid
OR9357-100G
OR9357-25G
OR9357-5G
APOSOR9357-100GEA
302
EUR
APOSOR9357-100G
APOSOR9357-25G
APOSOR9357-5G
2-Chloro-3-pyridineboronic acid 96%
2-Chloro-3-pyridineboronic acid
Formula:
C₅H₅BClNO₂ Melting Pt: 121 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094997 CAS Number: 381248-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...