5-Ethyl-2-thiophenecarboxaldehyde 98%

Supplier: Apollo Scientific

Synonyms: 5-Ethylthiophene-2-carbaldehyde

OR936277-25G OR936277-5G
APOSOR936277-25GEA 278 EUR
APOSOR936277-25G APOSOR936277-5G
5-Ethyl-2-thiophenecarboxaldehyde 98%
5-Ethyl-2-thiophenecarboxaldehyde
Formula: C₇H₈OS
MW: 140.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143457
CAS Number: 36880-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR