(±)-2-Phenyl-1-propanol 95%

Supplier: Apollo Scientific

Synonyms: 2-Phenylpropan-1-ol

OR936303-25G OR936303-500G
APOSOR936303-25GEA 65.5 EUR
APOSOR936303-25G APOSOR936303-500G
(±)-2-Phenyl-1-propanol 95%
(±)-2-Phenyl-1-propanol
Formula: C₆H₅CH(CH₃)CH₂OH
MW: 136,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004736
CAS Number: 1123-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR