N,N-Diethyl-1-naphthylamine 95%

Supplier: Apollo Scientific

OR936375-1G OR936375-5G
APOSOR936375-1GEA 285 EUR
APOSOR936375-1G APOSOR936375-5G
N,N-Diethyl-1-naphthylamine 95%
N,N-Diethyl-1-naphthylamine
Formula: C₁₄H₁₇N
MW: 199,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046389
CAS Number: 84-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR