N-Decanoylsarcosine sodium salt 98%
Supplier: Apollo Scientific
OR936414-1G
OR936414-5G
APOSOR936414-1GEA
174
EUR
APOSOR936414-1G
APOSOR936414-5G
N-Decanoylsarcosine sodium salt 98%
N-Decanoylsarcosine sodium salt
Formula:
C₁₃H₂₅NO₃ Melting Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00451533 CAS Number: 30377-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...