2-Phenylpropionitrile 95%

Supplier: Apollo Scientific

Synonyms: alpha-Methylphenylacetonitrile

OR936420-25G
APOSOR936420-25GEA 214 EUR
APOSOR936420-25G
2-Phenylpropionitrile 95%
2-Phenylpropionitrile
Formula: C₉H₉N
MW: 131,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001871
CAS Number: 1823-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR