Dimethylketene methyl trimethylsilyl acetal 95%

Supplier: Apollo Scientific

Synonyms: 1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, ((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane

OR936451-1G OR936451-25G OR936451-5G
APOSOR936451-1GEA 125 EUR
APOSOR936451-1G APOSOR936451-25G APOSOR936451-5G
Dimethylketene methyl trimethylsilyl acetal 95%
Dimethylketene methyl trimethylsilyl acetal
Formula: C₈H₁₈O₂Si
MW: 174,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010232
CAS Number: 31469-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR