3-Hydroxybenzophenone 97%
Supplier: Apollo Scientific
Synonyms:
m-Hydroxybenzophenone, (3-Hydroxyphenyl)(phenyl)methanone
OR936494-25G
OR936494-5G
APOSOR936494-25GEA
243
EUR
APOSOR936494-25G
APOSOR936494-5G
3-Hydroxybenzophenone 97%
3-Hydroxybenzophenone
Formula:
C₁₃H₁₀O₂ MW: 198,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002297 CAS Number: 13020-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...