3,3,5-Trimethylcaproic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3,3,5-Trimethylhexanoic acid, 3,5,5-Trimethylhexanoicacid, 3,5,5-Trimethylhexanoic acid

OR936533-100G OR936533-1KG OR936533-500G
APOSOR936533-100GEA 75.4 EUR
APOSOR936533-100G APOSOR936533-1KG APOSOR936533-500G
3,3,5-Trimethylcaproic acid 98%
3,3,5-Trimethylcaproic acid
Formula: C₉H₁₈O₂
MW: 158,24 g/mol
Boiling Pt: 120 °C/10mmHg
Melting Pt: –70 °C
Storage Temperature: Ambient
MDL Number: MFCD00020507
CAS Number: 3302-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR