p-Tolyl isovalerate 95%

Supplier: Apollo Scientific

Synonyms: p-Tolyl 3-methylbutanoate, Isovaleric Acid p-Tolyl Ester

OR936570-25G OR936570-5G OR936570-100G
APOSOR936570-25GEA 74.1 EUR
APOSOR936570-25G APOSOR936570-5G APOSOR936570-100G
p-Tolyl isovalerate 95%
p-Tolyl isovalerate
Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00036648
CAS Number: 55066-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR