4-Ethylphenyl acetate 95%

Supplier: Apollo Scientific

Synonyms: Acetic Acid 4-Ethylphenyl Ester

OR936654-25G OR936654-5G
APOSOR936654-25GEA 306 EUR
APOSOR936654-25G APOSOR936654-5G
4-Ethylphenyl acetate 95%
4-Ethylphenyl acetate
Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026970
CAS Number: 3245-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR