2-Phenylimidazoline 95%

Supplier: Apollo Scientific

Synonyms: 2-Phenyl-4,5-dihydro-1H-imidazole, 2-Phenyl-2-imidazoline

OR936688-25G OR936688-500G
APOSOR936688-25GEA 125 EUR
APOSOR936688-25G APOSOR936688-500G
2-Phenylimidazoline 95%
2-Phenylimidazoline
Formula: C₉H₁₀N₂
MW: 146,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005180
CAS Number: 936-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR