2-[(2-Methylphenoxy)methyl]oxirane 85%

Supplier: Apollo Scientific

Synonyms: 2-((o-Tolyloxy)methyl)oxirane

OR936726-100G OR936726-25G
APOSOR936726-100GEA 227 EUR
APOSOR936726-100G APOSOR936726-25G
2-[(2-Methylphenoxy)methyl]oxirane 85%
2-[(2-Methylphenoxy)methyl]oxirane
Formula: C₁₀H₁₂O₂
MW: 164,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037847
CAS Number: 2210-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR