Tiglic anhydride 95%
Supplier: Apollo Scientific
Synonyms:
(E)-2-Methyl-2-butenoic Anhydride, 2-Methylcrotonic Anhydride
OR936769-1G
OR936769-5G
APOSOR936769-1GEA
198
EUR
APOSOR936769-1G
APOSOR936769-5G
Tiglic anhydride 95%
Tiglic anhydride
Formula:
C₁₀H₁₄O₃ MW: 182,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00143614 CAS Number: 14316-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...