4-Butylresorcinol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Butylbenzene-1,3-diol

OR936805-1G OR936805-250MG
APOSOR936805-1GEA 95.7 EUR
APOSOR936805-1G APOSOR936805-250MG
4-Butylresorcinol 98%
4-Butylresorcinol
Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01684800
CAS Number: 18979-61-8
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR