N-Pentylaniline 95%

Supplier: Apollo Scientific

Synonyms: N-Amylaniline

OR936857-5G
APOSOR936857-5GEA 188 EUR
APOSOR936857-5G
N-Pentylaniline 95%
N-Pentylaniline
Formula: C₁₁H₁₇N
MW: 163,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059431
CAS Number: 2655-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR