Butyl-4-hydroxybenzoate 98%

Supplier: Apollo Scientific

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

OR937186-100G OR937186-1KG OR937186-500G
APOSOR937186-100GEA 125 EUR
APOSOR937186-100G APOSOR937186-1KG APOSOR937186-500G
Butyl-4-hydroxybenzoate 98%
Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194,23 g/mol
Boiling Pt: 156…157 °C/3,5mmHg
Melting Pt: 67…71 °C
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR