3-Acetylbenzo[b]thiophene 95%

Supplier: Apollo Scientific

Synonyms: 3-Acetylthianaphthene

OR937265-1G OR937265-5G
APOSOR937265-1GEA 154 EUR
APOSOR937265-1G APOSOR937265-5G
3-Acetylbenzo[b]thiophene 95%
3-Acetylbenzo[b]thiophene
Formula: C₁₀H₈OS
Storage Temperature: Ambient
MDL Number: MFCD00051638
CAS Number: 1128-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR