2-Ethyl-1-butanol 95%
Supplier: Apollo Scientific
OR937281-100G
OR937281-500G
APOSOR937281-100GEA
132
EUR
APOSOR937281-100G
APOSOR937281-500G
2-Ethyl-1-butanol 95%
2-Ethyl-1-butanol
Formula:
C₆H₁₄O MW: 102,18 g/mol Boiling Pt: 146 °C/760mmHg Melting Pt: –15 °C Density: 0,83 g/cm³ (25 °C) Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004744 CAS Number: 97-95-0 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...