N,N-Dimethylthioacetamide 95%
Supplier: Apollo Scientific
OR937293-1G
OR937293-5G
APOSOR937293-1GEA
37.7
EUR
APOSOR937293-1G
APOSOR937293-5G
N,N-Dimethylthioacetamide 95%
N,N-Dimethylthioacetamide
Formula:
C₄H₉NS MW: 103.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022178 CAS Number: 631-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...