2-Boc-2-azabicyclo[2.2.1]hept-5-ene 95%

Supplier: Apollo Scientific

Synonyms: tert-Butyl 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylate, 2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]hept-5-ene, 2-Azabicyclo[2.2.1]hept-5-ene-2-carboxylic acid tert-Butyl ester, 2-Boc-2-azabicyclo[2.2.1]hept-5-ene

OR937331-1G OR937331-250MG
APOSOR937331-1GEA 408 EUR
APOSOR937331-1G APOSOR937331-250MG
2-Boc-2-azabicyclo[2.2.1]hept-5-ene 95%
2-Boc-2-azabicyclo[2.2.1]hept-5-ene
Formula: C₁₁H₁₇NO₂
MW: 195,262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22416354
CAS Number: 188345-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR