1-Penten-3-ylacetate 95%
Supplier: Apollo Scientific
OR937491-10G
OR937491-1G
OR937491-5G
APOSOR937491-10GEA
770
EUR
APOSOR937491-10G
APOSOR937491-1G
APOSOR937491-5G
1-Penten-3-ylacetate 95%
1-Penten-3-ylacetate
Formula:
C₇H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02093427 CAS Number: 10500-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...