2-Amino-3'-chloroacetophenone hydrochloride 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chlorophenacylamine hydrochloride

OR9375-1G OR9375-5G
APOSOR9375-1GEA 216 EUR
APOSOR9375-1G APOSOR9375-5G
2-Amino-3'-chloroacetophenone hydrochloride 97%
2-Amino-3'-chloroacetophenone hydrochloride
Formula: C₈H₉Cl₂NO
Storage Temperature: Ambient
CAS Number: 51084-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR