Dimethyl citraconate 95%

Supplier: Apollo Scientific

Synonyms: Citraconic Acid Dimethyl Ester, (2Z)-Dimethyl 2-methylmaleate

OR937568-25G OR937568-5G
APOSOR937568-25GEA 203 EUR
APOSOR937568-25G APOSOR937568-5G
Dimethyl citraconate 95%
Dimethyl citraconate
Formula: C₇H₁₀O₄
MW: 158,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059306
CAS Number: 617-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR