(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 95%
Supplier: Apollo Scientific
Synonyms:
(1S)-1-(2-chlorophenyl)ethane-1,2-diol
OR937676-1G
OR937676-5G
APOSOR937676-1GEA
62.6
EUR
APOSOR937676-1G
APOSOR937676-5G
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol 95%
(S)-(+)-1-(2-Chlorophenyl)-1,2-ethanediol
Formula:
C₈H₉ClO₂ MW: 172,608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863241 CAS Number: 133082-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...